Learn More
PRECISION CELL SYSTEMS REFURBISHED MOXI GO II CELL
Supplier:  PRECISION CELL SYSTEMS MXG102RMR202503141245
Encompass_Preferred
  • Compact design for easy benchtop accessibility
  • Delivers rapid results in 5-10 seconds per sample
  • Maintenance-free operation with disposable cassettes
  • Utilizes Coulter principle for precise cell counting
  • Supports diverse applications including cell viability and genomics
  • Intuitive touchscreen interface for user-friendly operation
  • Portable with a Lithium Ion battery
  • Two-channel fluorescence detection capabilities
  • High particle size resolution down to 0.1 µm
  • USB connectivity for easy data transfer
Catalog No. NC3641944
Add to Cart
Request a Quote