Learn More
V&P Scientific Inc REGISTRATION BASE, High Profile
Supplier:  V&P Scientific Inc VP 771G4RM
Encompass
REGISTRATION BASE, SLAS Footprint, Black Anodized Aluminum Magnet Frame, High Profile, Robot, and Manual Aspiration and Dispensing Applications
Catalog No. NC1931693