Learn More
Medchemexpress LLC REG1A Cynomolgus H 5ug | 5ug
SDP
Supplier:  Medchemexpress LLC HYP774735UG
Encompass_Preferred
REG1A Protein serves as an inhibitor of spontaneous calcium carbonate precipitation REG1A Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived REG1A protein expressed by HEK293 with C-His labeled tag REG1A Protein Cynomolgus (HEK293 His) has molecular weight of 19 21 kDa respectively
Catalog No. 50-001-86743