Learn More
STEMCELL Technologies Human Frozen Bone Marrow Proficiency Testing Program - 1st Participant, Spring, Size: 1 Course
SDP
Supplier:  STEMCELL Technologies 1000926
Encompass
The Proficiency Testing program is designed to standardize the hematopoietic colony-forming unit (CFU) assay. Participants will be assessed on their proficiency at performing bone marrow colony assays using human bone marrow, focusing on the following aspects:1. Sample Preparation2. Plating and Set-Up of Colony CFU Assay3. Colony Enumeration and IdentificationProficiency Testing programs are available only in certain territories. Please contact your Sales representative or proficiency@stemcell.com for more information.
Catalog No. 50-226-9619