Learn More
Selleck Chemical LLC Regorafenib S1178-1g
SDP
Supplier:  Selleck Chemical LLC S11781G
Encompass_Preferred
Regorafenib is a multi-target inhibitor for VEGFR1 VEGFR2 VEGFR3 PDGFR Kit (c-Kit) RET (c-RET) and Raf-1 with IC50 of 13 nM/4 2 nM/46 nM 22 nM 7 nM 1 5 nM and 2 5 nM in cell-free assays respectively Regorafenib induces autophagy
Catalog No. 50-310-8274