Learn More
Medchemexpress LLC Regorafenib-d3 (BAY 73-4506-d3) | 1255386-16-3 | MFCD22380808 | 98.0% | 485.83 g·mol⁻¹ | C21H12D3ClF4N4O3 | 10 MG

Supplier: Medchemexpress LLC HY10331S10MG
Regorafenib-d3 is a deuterium-labeled analog of regorafenib supplied for use as a tracer and internal standard in quantitative analytical methods including LC-MS, GC-MS, and NMR. It is provided as a powder designed for method development, isotope-dilution assays, and pharmacokinetic studies.
- Deuterium-labeled internal standard for quantitative analysis
- Suitable for LC-MS, GC-MS, and NMR applications
- Supplied as a 10 mg powder
- High chemical purity (98.0%)
- Molecular weight 485.83 g·mol⁻¹; molecular formula C21H12D3ClF4N4O3
- Recommended storage: powder -20°C; in solvent -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.