Learn More
Cayman Chemical RegorafenIb N-oxIde 25mg
SDP
Supplier:  Cayman Chemical 3510125 MG
Encompass_Preferred
An active metabolite of regorafenib; formed from regorafenib by CYP3A4; inhibits VEGFR2, Tie2, c-Kit, and B-RAF in vitro; inhibits tumor growth in HT-29 and MDA-MB-231 mouse xenograft models at 1 mg/kg
Catalog No. 50-291-0646