Learn More
Medchemexpress LLC Rehmannioside C | 81720-07-2 | 1 MG

Supplier: Medchemexpress LLC HYN24001MG
Rehmannioside C is an iridoid glucoside isolated from Radix Rehmanniae Praeparata. It is intended for research use only.
- Purity of 99.47%
- Solid form, white to light yellow color
- Isolated from plants, specifically Scrophulariaceae and Rehmannia glutinosa
- Soluble in DMSO (50 mg/mL)
- Suitable for in vivo dissolution protocols
- Classified as a natural product standard, terpenoid, and iridoid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.