Learn More
Medchemexpress LLC Rel-HDMP 28 hydrochloride | 219915-69-2 | 99.7% | 319.83 | C18H22ClNO2 | 5mg
SDP
Supplier:  Medchemexpress LLC HYW710914A5MG
Encompass_Preferred
rel-HDMP 28 hydrochloride (Compound 2g) is a methylphenidate analogue that binds selectively to SERT (Ki 105 nM)[1]
Catalog No. 50-002-70058