Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (rel)-PROTAC ERRα degrader-1 | 00-00-0 | 98.2% | 1108.92 g/mol | C54H49Cl2F6N7O8 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY128838A5MG

Encompass_Preferred

(rel)-PROTAC ERRα Degrader-1 is a proteolysis-targeting chimera designed to induce degradation of estrogen-related receptor alpha (ERRα). The molecule combines an MDM2 ligand, a linker, and an ERRα-binding moiety and is supplied as a solid research reagent for laboratory studies.

  • Induces degradation of estrogen-related receptor alpha (ERRα).
  • PROTAC mechanism combining an MDM2 ligand and an ERRα-binding moiety.
  • Molecular weight 1108.92 g/mol.
  • Purity 98.18%.
  • Solid form, off-white to light yellow.
  • Supplied as a 5 mg research-use-only sample.

Catalog No. 50-002-62837


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.