Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC rel-Sertraline-d3 hydrochloride | 1330180-66-9 | 99.5% | 1 MG
SDP

Supplier:  Medchemexpress LLC HYB0176AS1MG

Encompass_Preferred

rel-Sertraline-d3 hydrochloride is a deuterium-labeled form of Sertraline hydrochloride, an antidepressant of the selective serotonin reuptake inhibitor (SSRI) class. It is utilized in research for various diseases, including major depressive disorder. This compound serves as a tracer or an internal standard for quantitative analysis. Its molecular weight is 345.71 and its formula is C17H15D3Cl3N. It appears as a solid and ships at room temperature within the continental US.

  • Deuterium-labeled Sertraline hydrochloride
  • Antidepressant of the selective serotonin reuptake inhibitor (SSRI) class
  • Used as a tracer or internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Molecular weight of 345.71
  • Chemical formula C17H15D3Cl3N
  • Long-term powder storage at -20°C for 3 years
  • Solution storage at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-05998


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.