Learn More
Medchemexpress LLC (rel)-Tivantinib ((rel)-ARQ 197) | 905853-99-8 | 96.45% | C23H19N3O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY7749350MG
Encompass_Preferred
(rel)-Tivantinib, also known as (rel)-ARQ 197, is a potent and highly selective inhibitor of the receptor tyrosine kinase c-MET. This compound is known for its role in inhibiting cancer cell growth and proliferation by targeting the c-MET pathway, which is often dysregulated in various cancers. It has been investigated for its potential as an anti-cancer agent due to its ability to block c-MET signaling.
  • Potent and highly selective c-MET inhibitor.
  • Inhibits cancer cell growth and proliferation.
  • Targets the c-MET signaling pathway.
  • Investigated as a potential anti-cancer agent.
Catalog No. 50-004-34939