Learn More
Medchemexpress LLC Relamorelin TFA | 2863659-22-5 | 99.8% | 905.00 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY19884B5MG
Encompass_Preferred
Relamorelin TFA, a pentapeptide ghrelin analog, is a selective ghrelin/growth hormone secretagogue receptor (GHSR) agonist with a Ki of 0.42 nM for GHS-1a receptor. It is centrally penetrant, increases growth hormone levels, and accelerates gastric emptying.
  • Selective ghrelin/growth hormone secretagogue receptor (GHSR) agonist
  • Centrally penetrant
  • Increases growth hormone levels
  • Accelerates gastric emptying
  • Potential for cachexia research
  • Potential for gastroparesis research
  • Potential for gastric/intestinal dysmobility disorders research
Catalog No. 50-004-03324