Learn More
Grainger Relay 120VAC
Supplier:  Grainger 16X422
Encompass_Preferred
The Edwards Signaling 120V AC Relay is a general purpose relay that can be used for a variety of signaling and control applications. It has a 120V AC coil voltage and 10 amp contacts rated for 120V AC. The relay has an octal plug with screw terminals for easy wiring. Some key features include the dust cover for protection, clear polycarbonate cover for visibility, and quick-connect socket base. This industrial-grade relay is UL/CSA recognized and manufactured in the USA. With its versatile design, it can provide reliable performance for controlling various devices and circuits.
Catalog No. 50-276-7544