Learn More
TARGETMOL CHEMICALS INC RELEBACTAM 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T401710MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Relebactam (MK-7655) is a diazabicyclooctane inhibitor with activity against a wide spectrum of (beta)-lactamases. purity: 100%
Catalog No. 50-258-0001