Learn More
Selleck Chemical LLC Remdesivir S8932-5MG
SDP
Supplier:  Selleck Chemical LLC S89325MG
Encompass_Preferred
Remdesivir (GS-5734) a monophosphoramidate prodrug of an adenosine analog is an investigational broad-spectrum antiviral agent with in vitro activity against multiple RNA viruses including Ebola and CoV
Catalog No. 50-000-23650