Learn More
Medchemexpress LLC Remdesivir triphosphate trisodium | 1355050-21-3 | 100.0% | 597.15 | C12H13N5Na3O13P3 | 100MG
SDP
Supplier:  Medchemexpress LLC HY126303C100MG
Encompass_Preferred
GS-443902 trisodium is the triphosphate metabolite of remdesivir supplied for research use. It acts as an adenosine triphosphate analogue and inhibits viral RNA-dependent RNA polymerases, making it useful for antiviral mechanism studies and metabolite profiling.
  • Active triphosphate metabolite of remdesivir.
  • Inhibits viral RNA-dependent RNA polymerase (RdRp).
  • Supplied as a trisodium salt solid for research use.
  • High reported purity (99.98% by RP-HPLC).
  • Available in multiple pack sizes suitable for small-scale experiments.
Catalog No. 50-002-37379
Add to Cart
Request a Quote