Learn More
Medchemexpress LLC HY-128757 10mg , Remibrutinib CAS:1787294-07-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12875710MG
Encompass_Preferred
HY-128757 10mg Remibrutinib CAS: 1787294-07-8 Remibrutinib, is a potent and orally active bruton tyrosine kinase (BTK) inhibitor with an IC50 value of 1 nM. Remibrutinib inhibits BTK activity with an IC50 value of 0.023 μM in blood. Remibrutinib has the potential for Chronic urticaria (CU) treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0668