Learn More
Medchemexpress LLC Remibrutinib | 1787294-07-8 | 99.8% | 1 MG

Supplier: Medchemexpress LLC HY1287571MG
Remibrutinib is a potent and orally active Bruton Tyrosine Kinase (BTK) inhibitor with an IC50 value of 1 nM. It inhibits BTK activity with an IC50 value of 0.023 μM in blood and has potential for the treatment of Chronic Urticaria (CU).
- Potent and orally active BTK inhibitor
- Inhibits BTK activity with an IC50 of 1 nM
- Has potential for chronic urticaria treatment
- Purity of 99.8%
- Molecular weight of 507.53
- Appears as a white to off-white solid
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.