Learn More
Medchemexpress LLC Remibrutinib | 1787294-07-8 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12875710MM/1ML
Encompass_Preferred
Remibrutinib is a potent and orally active Bruton tyrosine kinase (BTK) inhibitor with an IC50 value of 1 nM. It inhibits BTK activity with an IC50 value of 0.023 μM in blood. This compound has the potential for Chronic Urticaria (CU) treatment.
  • Potent and orally active Bruton tyrosine kinase (BTK) inhibitor
  • Inhibits BTK activity with an IC50 value of 1 nM
  • Inhibits BTK activity in blood with an IC50 value of 0.023 μM
  • Potential for Chronic Urticaria (CU) treatment
Catalog No. 50-003-39640