Learn More
Medchemexpress LLC Remibrutinib | 1787294-07-8 | 99.8% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12875750MG
Encompass_Preferred
Remibrutinib is a potent and orally active Bruton tyrosine kinase (BTK) inhibitor. It inhibits BTK activity with an IC50 value of 1 nM and 0.023 μM in blood, showing potential for chronic urticaria (CU) treatment.
  • Potent and orally active Bruton tyrosine kinase (BTK) inhibitor.
  • Inhibits BTK enzymatic activity at 1 nM.
  • Effective at inhibiting BTK activity in blood at 0.023 μM.
  • Shows promise for the treatment of chronic urticaria.
Catalog No. 50-003-42848