Learn More
Medchemexpress LLC Remogliflozin etabonate | 442201-24-3 | MFCD17167013 | 99.8% | 522.59 | C26H38N2O9 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1494510MG
Encompass_Preferred
Remogliflozin etabonate is an orally active prodrug that is metabolized to remogliflozin, a selective sodium-glucose cotransporter 2 (SGLT2) inhibitor used in preclinical antidiabetic research. In rodent studies it increases urinary glucose excretion, lowers fasting plasma glucose and glycated hemoglobin, and can suppress hypertriglyceridemia.
  • Selective SGLT2 inhibition for antidiabetic research.
  • Orally active prodrug metabolized to the active glucoside.
  • High purity (~99.8%) for reproducible experimental results.
  • White to off-white solid, suitable for handling and storage.
  • Available in small laboratory quantities for in vitro and in vivo studies.
  • Documented efficacy in multiple rodent models.
Catalog No. 50-002-85914