Learn More
Medchemexpress LLC Remogliflozin etabonate (GSK189075) | 442201-24-3 | 99.64% | 522.59 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY149455MG
Encompass_Preferred
Remogliflozin etabonate (GSK189075) is an orally active, selective, and low-affinity sodium glucose cotransporter (SGLT2) inhibitor. It functions as a proagent derived from benzylpyrazole glucoside and is metabolized into its active form, Remogliflozin, within the body. This compound has demonstrated antidiabetic efficacy in rodent models.
  • Orally active and selective SGLT2 inhibitor
  • Functions as a proagent of Remogliflozin
  • Exhibits antidiabetic efficacy in rodent models
Catalog No. 50-003-60314