Learn More
Grainger Remote Test Station 2 Watt
Supplier:  Grainger 16X440
Encompass_Preferred
The Edwards Signaling Remote Test Station is a 2-watt remote test station that allows for testing of fire alarm systems. It features a red ABS plastic enclosure and includes a key operated test switch and alarm LED indicator. The test station has screw terminal connections and operates on 24VDC. It provides a simple method to manually activate and test notification appliances in a fire alarm system. The compact design can be easily installed where needed for periodic system testing.
Catalog No. 50-276-6969