Learn More
GA International Removable Autoclave-Resistant Thermal-Transfer Labels 1.25" x 0.875" / 31.8mm x 22.2mm Rectangle White From -60°C to +150°C (-76°F to +302°F)
Supplier:  GA International AUTR9C33WH
Encompass_Preferred
Removable autoclave-resistant labels, ideal for metal, plastic, and glass surfaces, including surgical instruments, glassware, trays, blister packs, and other items intended for use in steam autoclaves, ovens, gamma irradiators, and ethylene oxide sterilization. Our durable autoclave thermal-transfer labels will not leave any adhesive residue after removal. They are weather & water-proof, alcohol-resistant, and have been tested to endure up to 20 autoclave cycles (at 121°C/250°F, 16.5 psi for 20 minutes). Compatible with all major brands of thermal-transfer printers.The upper-temperature point might slightly vary depending on the duration of the exposure.*Not recommended for dishwasher use as ink may smudge when exposed to detergents. For dishwasher-resistant labels, please contact us.
Catalog No. 50-005-71774