Learn More
GA International Removable Cryogenic Labels for Laser Printers 1.42" x 0.55" / 36.1mm x 14mm Rectangle Purple From -196°C to +120°C (-321°F to +248°F)
Supplier:  GA International RCL6T1PU
Encompass_Preferred
Cryogenic thermoplastic laser labels with a glove-friendly removable adhesive. Ideal for long-term storage of samples in liquid nitrogen (on flat surfaces such as polycarbonate cryo boxes) and harsh deep-freeze environments (as low as -80°C; on vials, tubes, freezer boxes, microplates, and others) used in biomedical research and clinical laboratories. Cryo labels accept desktop laser printing and permanent markers. Water-resistant; wear and tear proof; do not jam printers; resistant to UV and gamma irradiation.
Catalog No. 50-005-70889