Learn More
GA International Removable Deep-Freeze Labels for Thermal-Transfer Barcode Printers 3" x 1.625" / 76.2mm x 41.3mm Rectangle White From -80°C to +100°C (-112°F to +212°F)
Supplier:  GA International RMTT533NPC31WH
Encompass_Preferred
Deep-freeze, removable barcode labels for vials, tubes, as well as polypropylene and cardboard boxes. The freezer labels have a strong, glove-friendly adhesive that allows them to adhere firmly to containers in (-20°C, -40°C, -80°C) freezers and dry ice while still being easily removable at room temperature without leaving behind any residue.Waterproof and tearproof, these removable deep-freeze labels can also withstand temperatures ranging up to +100°C, making them suitable for general lab use such as the identification of beakers, flasks, boxes, plastic and glass bottles that are stored in fridges and freezers.
Catalog No. 50-005-71140