Learn More
Cayman Chemical RemoxIprIdehydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 309715 MG
Encompass_Preferred
An atypical antipsychotic and dopamine D2 receptor antagonist (Ki = 0.2 µM); selective for the D2 receptor over a panel of 33 receptors, neurotransmitter transporters, and ion channels at 10 µM but does inhibit the σ receptor (Ki = 0.065 µM); increases striatal DOPAC and HVA levels in rats from 2-20 µmol/kg; inhibits apomorphine-induced stereotypy and hyperactivity in rats (ED50s = 5.6 and 0.8 µmol/kg, respectively)
Catalog No. 50-298-9008