Learn More
ATCC Renal Epithelial Cell Basal Medium
Supplier:  ATCC PCS400030
Encompass
Renal Epithelial Cell Basal Medium is a sterile, phenol-red-free, liquid tissue culture medium intended for use as one component in a complete ATCC Primary Cell Solutions system. This system is designed to support epithelial cells (eg, renal proximal tubule epithelial cells) derived from a normal human kidney. Renal Epithelial Cell Basal Medium contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts.
Catalog No. 50-238-2400
 Restricted Item