Learn More
ATCC Renca; Renal Adenocarcinoma; Mouse (Mus musculus)
Supplier:  ATCC CRL2947
Encompass
Renca is an epithelial cell that was isolated from the kidney of a male mouse with renal cortical adenocarcinoma. The cell was deposited by R Wiltrout in 1969.
Catalog No. 50-238-3796