Learn More
MSC Renishaw CMM Ball Stylus, 0.70 mm Ruby, 10 mm OAL, M2
Supplier:  MSC 00552042
Encompass
This CMM ball stylus is designed for precise contact with workpiece surfaces in coordinate measuring machines. It features a durable ruby tip and a carbide stem, providing accuracy and longevity for various measurement applications.
  • Spherical tip for precise contact with workpiece surfaces.
  • Tip made of synthetic ruby.
  • Mounted on a carbide stem.
Catalog No. 50-005-29603