Learn More
MSC Renishaw CMM Ball Stylus, 3 mm Ball Dia, 50 mm OAL, M3 Thread
Supplier:  MSC 00552489
Encompass
This Renishaw CMM Ball Stylus is designed for coordinate measuring machines. It features a 3 mm ball diameter, a 50 mm overall length, and an M3 thread. The stylus utilizes a durable ruby point and a robust solid carbide stem, ensuring precision and longevity in various measurement applications.
  • 3 mm ball diameter
  • 50 mm overall length
  • M3 thread size
  • 43.70 mm effective working length
  • Ruby point material
  • Solid carbide stem material
  • 2.00 mm stem diameter
Catalog No. 50-005-30031