Learn More
Medchemexpress LLC Repaglinide-d5 | 1217709-85-7 | MFCD08063596 | 98.7% | 457.62 | C27H31D5N2O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15209S10MG
Encompass_Preferred
Repaglinide-d5 is a deuterium-labeled analogue of repaglinide supplied for research and analytical use. It is intended as an internal standard or tracer for quantitative analysis and method development, and is used to support studies of repaglinide pharmacology and metabolism.
  • Deuterium-labeled internal standard for analytical quantification.
  • Suitable for LC-MS, GC-MS, and NMR applications.
  • High purity to support accurate analytical results.
  • Provided as a small research-grade sample for method validation and tracer studies.
Catalog No. 50-002-85896