Learn More
Medchemexpress LLC Reparixin | 266359-83-5 | C14H21NO3S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15251A50MG
Encompass_Preferred
Reparixin is a non-competitive allosteric inhibitor that targets the chemokine receptors CXCR1 and CXCR2. It is a potent functional inhibitor of CXCL8-induced biological activities and has been shown to attenuate inflammatory responses in various injury models. This compound is for research use only.
  • Non-competitive allosteric inhibitor of CXCR1 and CXCR2 activation
  • Potent inhibitor of CXCL8-induced biological activities
  • Attenuates inflammatory responses
  • Demonstrated to decrease systolic blood pressure and increase blood flow in animal models
  • Reduces IL-1β levels in the brain in animal models
Catalog No. 50-003-73655