Learn More
Medchemexpress LLC Reparixin L-lysine salt | 266359-93-7 | C20H35N3O5S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1525210MM/1ML
Encompass_Preferred
Reparixin L-lysine salt is an allosteric inhibitor of chemokine receptor 1/2 (CXCR1/2) activation. It is for research use only and not sold to patients. It is a potent functional inhibitor of CXCL8-induced biological activities on human PMNs with marked selectivity (around 400-fold) for CXCR1. Reparixin is a non-competitive allosteric inhibitor of IL-8 receptors with a 400-fold higher efficacy in inhibiting CXCR1 activity than CXCR2.
  • Allosteric inhibitor of chemokine receptor 1/2 (CXCR1/2) activation.
  • Potent functional inhibitor of CXCL8-induced biological activities on human PMNs.
  • Marked selectivity (around 400-fold) for CXCR1.
  • Non-competitive allosteric inhibitor of IL-8 receptors.
  • 400-fold higher efficacy in inhibiting CXCR1 activity than CXCR2.
  • For research use only.
Catalog No. 50-003-73656