Learn More
USA Scientific Inc Repeater E3x with power cable
Supplier:  USA Scientific Inc 40497134
Encompass
Eppendorf Repeater E3x, 1-channel, with charging cable and Combitips advanced assortment pack (1 Combitips advanced of each size), 1 uL-50 mL
Catalog No. 50-274-4986