Learn More
Cole-Parmer Replacement glass beads, 2 mm dia., 10 oz per bag
Supplier:  Cole-Parmer 1077950
Encompass
Keep your sterilizer working at its best with these parts and accessories. These replacement glass beads for dry glass bead sterilizers are two mm in diameter. A10 ounce bag refills the three inch diameter sterilizer twice, and the five inch diameter sterilizer once.
Catalog No. NC9566553
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.