Learn More
Leica Biosystems Bacti-Cinerator IV Heating Element, 120V - Leica Biosystems (MPN 39001373)
Supplier:  Leica Biosystems 39001373
Encompass_Preferred
This replacement heating element is designed for use in the Leica Bacti-Cinerator IV heat chamber to restore infrared heating and rapid sterilization performance. It installs in the device's heat chamber and enables effective sterilization of inoculation loops and needles.
  • Replacement ceramic-core heating element for infrared sterilization.
  • Operates at 120 V for compatibility with 120 V Bacti-Cinerator IV units.
  • Enables rapid sterilization of loops and needles, minimizing spatter and downtime.
  • Restores heat chamber performance to factory heating characteristics.
  • No manufacturer dimensions or weight are listed; verify compatibility before ordering.
Catalog No. NC3443140