Learn More
Chemglass Life Sciences Chemglass Replacement 19 mm Stirrer Shaft for 10/15/20 L Filter Reactors
SDP
Supplier:  Chemglass Life Sciences CG209716
Encompass_Preferred
Replacement 19 mm stirrer shaft designed for process filter reactors. Precision ground and polished with a sandblasted lower section, this shaft restores reliable mixing performance in 10 L, 15 L, and 20 L reactors.
  • Precision ground and polished for smooth rotation.
  • 19 mm outside diameter.
  • Overall length 32.75 in.
  • Sandblasted lower portion with 3/8 in wide flat for secure blade mounting.
  • Fits 10 L, 15 L, and 20 L filter reactors and compatible support frames.
  • Replacement part to restore mixing performance in process reactors.
Catalog No. 50-273-3532