Learn More
Research Products International Corp Replacement Cover for Immuno-Staining Tray, Clear
SDP
Supplier:  Research Products International Corp 195802
Encompass
Replacement cover for use with Immuno-Staining tray. Clear replacement cover allows for visual examination. Made of PETG with a temperature range of -20 °C to +60 °C.
Catalog No. 50-197-7985