Learn More
Chemglass Life Sciences Replacement Glass Shaft, 500mL Internal Spinner Flask
SDP
Supplier:  Chemglass Life Sciences CLS142621
Encompass_Preferred
Replacement components for CLS-1414 and CLS-1415 Spinner Flasks.The Complete Impeller Assembly consists of top cap, silicone liner, glass rod, compression fitting magnet holder, paddle and magnetic stir bar.
Catalog No. 50-273-3296