Learn More
Chemglass Life Sciences Chemglass replacement S.S. retention kit for CG-468 stopcocks, plug size 16.2/56 (CG-469-23)
SDP
Supplier:  Chemglass Life Sciences CG46923
Encompass_Preferred
Replacement stainless-steel retention kit for Chemglass high-vacuum glass stopcocks. Provides the constant-pressure retention device components used to secure glass plugs in high-vacuum stopcock assemblies and is intended for maintenance and repair.
  • Provides stainless-steel retention components for the constant-pressure retention device.
  • Designed for plug size 16.2/56.
  • Intended for maintenance and repair of high-vacuum glass stopcocks.
  • Sold as a single kit (1 each) for straightforward replacement.
Catalog No. 50-273-3525