Learn More
Chemglass Life Sciences Replacement Sparger Tip for Air Aeration Tubes, 10 μm
SDP
Supplier:  Chemglass Life Sciences CLS138014
Encompass
Replacement sparger tip designed for air aeration tubes used in bioreactors. The porous stainless steel tip (10 μm) delivers fine bubble aeration for laboratory bioprocessing and is sold individually.
  • Replacement tip for air aeration tubes used in bioreactors.
  • Porosity of 10 μm for fine bubble aeration.
  • Porous stainless steel construction.
  • Sold individually (each).
  • Provides fine bubble dispersion for efficient gas transfer.
Catalog No. 50-273-3326