Learn More
REPLIGEN CORPORATION REPLIGEN A-PROTEIN ELISA KIT
SDP
Supplier:  REPLIGEN CORPORATION 90001
Encompass
Repligen Protein A ELISA for the detection of native and recombinant Protein A. Used for downstream processes which utilize a Protein A capture step (non-MabSelect SuRe™), Repligen offers the Protein A ELISA Kit suitable for Protein A resins that contain native and recombinant Protein A ligand variants. Includes 3 sample prep methods, Dilute and Go (no boiling or dialysis), Boil & Boost (low conc. samples), and Original.
Catalog No. NC0132803