Learn More
Medchemexpress LLC HY-103022 10mg Medchemexpress, Repotrectinib CAS:1802220-02-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10302210MG
Encompass_Preferred
Medchemexpress, HY-103022 10mg Repotrectinib CAS:1802220-02-5 Repotrectinib (TPX-0005) is a potent ROS1 (IC50=0.07 nM) and TRK (IC50=0.83/0.05/0.1 nM for TRKA/B/C) inhibitor. Repotrectinib potently inhibits WT ALK (IC50=1.01 nM). Repotrectinib has anti-cancer activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8722