Learn More
Apexbio Technology LLC Repotrectinib 1802220-02-5 10mg
SDP
Supplier:  Apexbio Technology LLC A465010
Encompass_Preferred
Repotrectinib (CAS 1802220-02-5) is a macrocyclic small molecule inhibitor of tyrosine kinases developed to overcome resistance mechanisms in non-small cell lung cancer (NSCLC) mediated by ROS1 gene mutations Its compact structure enables selective inhibition of both wild-type and solvent-front mutated ROS1 including the G2032R variant frequently associated with resistance to first-generation TKIs such as crizotinib Repotrectinib has demonstrated robust clinical activity in the TRIDENT-1 study with an objective response rate of 79% in TKI-naive and 38% in TKI-pretreated ROS1-positive NSCLC patients It serves as a valuable tool for investigating mechanisms of kinase inhibitor resistance in oncogenic ROS1-driven cancers
Catalog No. 50-000-93502