Learn More
Selleck Chemical LLC Repotrectinib (TPX-0005) 5mg 1802220-02-5
SDP
Supplier:  Selleck Chemical LLC S85835MG
Encompass_Preferred
Repotrectinib (TPX-0005) is a novel ALK/ROS1/TRK inhibitor with the IC50 values of 1.01 nM for WT ALK, 1.26 nM for ALK(G1202R), and 1.08 nM for ALK(L1196M); also a potent SRC inhibitor (IC50 5.3 nM). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8122