Learn More
Medchemexpress LLC Resazurin sodium | 62758-13-8 | 98.15% | 251.17 | 1 ML
Supplier: Medchemexpress LLC HY111391R100MG
Resazurin (sodium) (Standard) is the analytical standard of Resazurin (sodium) and is intended for research and analytical applications. Resazurin sodium (Diazoresorcinol sodium) is commonly used to measure bacterial and eukaryotic cell viability through its reduction to the fluorescent product resorufin.
- The compound is the grade of analytical standard, which is the reference standard supplied assay.
- Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.