Learn More
ACROBiosystems resDetect Kanamycin ELISA Kit
Supplier:  ACROBiosystems RESA00496TESTS
Encompass_Preferred
Residues of kanamycin are prone to occur in the production process of biological products which can easily lead to abnormal reactions in the human body. Therefore the residual amount of kanamycin in biological products or semi-finished products of biological products should be strictly controlled.
Catalog No. 50-253-4250