Learn More
Cell Sciences Inc Research Grade Alnuctamab, Trivalent Bispecific Monoclonal Antibody, 1.0 mg
Supplier:  Cell Sciences Inc DHF92403B
Encompass
Research Grade Alnuctamab is a trivalent, bispecific humanized monoclonal antibody designed for laboratory research applications. It uniquely targets human BCMA and CD3e, facilitating T-cell-mediated targeting, making it an advanced reagent for enhanced research results. Provided in a quantity of 1.0 mg, it ensures optimal performance across various research protocols, delivering stability for up to 1 year.
Catalog No. NC3198473